1-(4-Ethoxyphenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039845-500MG
101848-306EA 217.35 USD
101848-306
1-(4-Ethoxyphenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-(4-Ethoxyphenyl)-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₉H₂₁ClN₂O
MDL Number: MFCD07626285
CAS Number: 367251-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR