1-[4-(Trifluoromethyl)phenyl]-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039844-500MG
101848-304EA 290.77 USD
101848-304
1-[4-(Trifluoromethyl)phenyl]-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-[4-(Trifluoromethyl)phenyl]-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₈H₁₆ClF₃N₂
MW: 352.79 g/mol
MDL Number: MFCD07626283
CAS Number: 367251-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR