1-Phenyl-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039842-500MG
101848-300EA 193.85 USD
101848-300
1-Phenyl-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-Phenyl-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₇ClN₂
MW: 284.79 g/mol
Melting Pt: 254…258 °C
MDL Number: MFCD01690343
CAS Number: 3574-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR