2-Chloro-4-nitro-1H-imidazole ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-nitro-1H-imidazole

039841-1G 039841-5G
101848-296EA 49.23 USD
101848-296 101848-298
2-Chloro-4-nitro-1H-imidazole ≥97%
2-Chloro-4-nitro-1H-imidazole

Hazard: Irritant

Formula: C₃H₂ClN₃O₂
MW: 147.518 g/mol
MDL Number: MFCD03419295
CAS Number: 57531-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR