(3Z)-5-Chloro-3-(phenylimino)-1,3-dihydro-2H-indol-2-one

Supplier: Matrix Scientific

039834-500MG
101848-284EA 217.35 USD
101848-284
(3Z)-5-Chloro-3-(phenylimino)-1,3-dihydro-2H-indol-2-one
(3Z)-5-Chloro-3-(phenylimino)-1,3-dihydro-2H-indol-2-one

Hazard: Irritant

Formula: C₁₄H₉ClN₂O
MDL Number: MFCD12028076
CAS Number: 60283-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR