4,6-Dimethyl-2-thioxo-1,2-dihydropyridine-3-carbonitrile

Supplier: Matrix Scientific

039832-500MG
101848-280EA 290.77 USD
101848-280
4,6-Dimethyl-2-thioxo-1,2-dihydropyridine-3-carbonitrile
4,6-Dimethyl-2-thioxo-1,2-dihydropyridine-3-carbonitrile

Hazard: Irritant

Formula: C₈H₈N₂S
MDL Number: MFCD00186534
CAS Number: 54585-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR