1,2,4-Thiadiazole-3,5-bis(thiolate)

Supplier: Matrix Scientific

039831-500MG
101848-278EA 144.9 USD
101848-278
1,2,4-Thiadiazole-3,5-bis(thiolate)
1,2,4-Thiadiazole-3,5-bis(thiolate)

Hazard: Irritant

Formula: C₂BaN₂S₃
MDL Number: MFCD12028075
CAS Number: 82935-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR