Sodium-4-cyanoisothiazole-3,5-bis(thiolate)

Supplier: Matrix Scientific

039830-500MG
101848-276EA 193.85 USD
101848-276
Sodium-4-cyanoisothiazole-3,5-bis(thiolate)
Sodium-4-cyanoisothiazole-3,5-bis(thiolate)

Hazard: Irritant

Formula: C₄H₈N₂Na₂O₄S₃
MW: 290.29 g/mol
MDL Number: MFCD12028074
CAS Number: 1314229-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR