4-(5-Methyl-1,2,4-oxadiazol-3-yl)-1,2,5-oxadiazol-3-amine

Supplier: Matrix Scientific

039827-500MG
101848-270EA 387.69 USD
101848-270
4-(5-Methyl-1,2,4-oxadiazol-3-yl)-1,2,5-oxadiazol-3-amine
4-(5-Methyl-1,2,4-oxadiazol-3-yl)-1,2,5-oxadiazol-3-amine

Hazard: Irritant

Formula: C₅H₅N₅O₂
MDL Number: MFCD00168686
CAS Number: 164363-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR