3-(3,4-Dimethoxyphenyl)-1H-pyrazole-4-carbonitrile

Supplier: Matrix Scientific

039826-500MG
101848-268EA 387.69 USD
101848-268
3-(3,4-Dimethoxyphenyl)-1H-pyrazole-4-carbonitrile
3-(3,4-Dimethoxyphenyl)-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₂
MDL Number: MFCD12028073
CAS Number: 1159697-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR