N-Benzyl-2-[(2-bromopyridin-3-yl)oxy]acetamide

Supplier: Matrix Scientific

039825-500MG
101848-266EA 387.69 USD
101848-266
N-Benzyl-2-[(2-bromopyridin-3-yl)oxy]acetamide
N-Benzyl-2-[(2-bromopyridin-3-yl)oxy]acetamide

Hazard: Irritant

Formula: C₁₄H₁₃BrN₂O₂
MDL Number: MFCD12028072
CAS Number: 596808-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR