(4-Nitrophenoxy)acetonitrile

Supplier: Matrix Scientific

Synonyms: 2-(4-Nitrophenoxy)acetonitrile

039823-500MG
101848-262EA 146.15 USD
101848-262
(4-Nitrophenoxy)acetonitrile
(4-Nitrophenoxy)acetonitrile

Hazard: Irritant

Formula: C₈H₆N₂O₃
MW: 178.1448 g/mol
MDL Number: MFCD00068120
CAS Number: 33901-46-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR