5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1-methyl-2-oxindole

039822-500MG
101848-260EA 144.9 USD
101848-260
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one
5-Bromo-1-methyl-1,3-dihydro-2H-indol-2-one

Hazard: Irritant

Formula: C₉H₈BrNO
MW: 226.08 g/mol
MDL Number: MFCD00464061
CAS Number: 20870-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR