3-[4-Formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]propanoic acid

Supplier: Matrix Scientific

039821-500MG
101848-258EA 387.69 USD
101848-258
3-[4-Formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₄
MDL Number: MFCD02725434
CAS Number: 381214-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR