2-(4-Aminophenoxy)-N-(4-methoxyphenyl)acetamide

Supplier: Matrix Scientific

039814-500MG
101848-244EA 217.35 USD
101848-244
2-(4-Aminophenoxy)-N-(4-methoxyphenyl)acetamide
2-(4-Aminophenoxy)-N-(4-methoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O₃
MDL Number: MFCD06662340
CAS Number: 857495-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR