Methyl-4-{2-[(3,4-dichlorophenyl)amino]-2-oxoethoxy}benzoate

Supplier: Matrix Scientific

039811-500MG
101848-234EA 290.77 USD
101848-234
Methyl-4-{2-[(3,4-dichlorophenyl)amino]-2-oxoethoxy}benzoate
Methyl-4-{2-[(3,4-dichlorophenyl)amino]-2-oxoethoxy}benzoate

Hazard: Irritant

Formula: C₁₆H₁₃Cl₂NO₄
MDL Number: MFCD06662076
CAS Number: 887358-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR