2-(4-Methylphenyl)-1,3-dioxoisoindoline-5-carbonyl chloride

Supplier: Matrix Scientific

039810-500MG
101848-232EA 193.85 USD
101848-232
2-(4-Methylphenyl)-1,3-dioxoisoindoline-5-carbonyl chloride
2-(4-Methylphenyl)-1,3-dioxoisoindoline-5-carbonyl chloride

Hazard: Irritant

Formula: C₁₆H₁₀ClNO₃
MDL Number: MFCD12028068
CAS Number: 1134335-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR