N,N'-Bis(2,4-diaminophenyl)terephthalamide
Supplier: Matrix Scientific
039809-500MG
101848-230EA
193.85
USD
101848-230
N,N'-Bis(2,4-diaminophenyl)terephthalamide
N,N'-Bis(2,4-diaminophenyl)terephthalamide
Hazard: Irritant
Formula:
C₂₀H₂₀N₆O₂ |
MDL Number:
MFCD00601721 CAS Number: 132663-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...