4-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]-1,2,5-oxadiazol-3-amine

Supplier: Matrix Scientific

039808-500MG
101848-228EA 289.8 USD
101848-228
4-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]-1,2,5-oxadiazol-3-amine
4-[5-(Chloromethyl)-1,2,4-oxadiazol-3-yl]-1,2,5-oxadiazol-3-amine

Hazard: Irritant

Formula: C₅H₄ClN₅O₂
MDL Number: MFCD01459277
CAS Number: 329212-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR