Methyl-4-[2-(cyclohexylamino)-2-oxoethoxy]benzoate

Supplier: Matrix Scientific

039803-500MG
101848-218EA 193.85 USD
101848-218
Methyl-4-[2-(cyclohexylamino)-2-oxoethoxy]benzoate
Methyl-4-[2-(cyclohexylamino)-2-oxoethoxy]benzoate

Hazard: Irritant

Formula: C₁₆H₂₁NO₄
MDL Number: MFCD08072234
CAS Number: 915880-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR