Furan-2,5-dicarbohydrazide

Supplier: Matrix Scientific

039801-500MG
101848-214EA 193.85 USD
101848-214
Furan-2,5-dicarbohydrazide
Furan-2,5-dicarbohydrazide

Hazard: Irritant

Formula: C₆H₈N₄O₃
MDL Number: MFCD00612475
CAS Number: 26095-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR