2-[(3,5-Dimethylphenyl)amino]-2-thioxoacetamide

Supplier: Matrix Scientific

039800-500MG
101848-212EA 144.9 USD
101848-212
2-[(3,5-Dimethylphenyl)amino]-2-thioxoacetamide
2-[(3,5-Dimethylphenyl)amino]-2-thioxoacetamide

Hazard: Irritant

Formula: C₁₀H₁₂N₂OS
MDL Number: MFCD01423557
CAS Number: 328961-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR