Methyl-4-[(2-cyanobenzyl)oxy]benzoate

Supplier: Matrix Scientific

039799-500MG
101848-210EA 290.77 USD
101848-210
Methyl-4-[(2-cyanobenzyl)oxy]benzoate
Methyl-4-[(2-cyanobenzyl)oxy]benzoate

Hazard: Irritant

Formula: C₁₆H₁₃NO₃
MDL Number: MFCD12028067
CAS Number: 1004249-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR