2-(2-Benzoylhydrazino)-N-phenyl-2-thioxoacetamide

Supplier: Matrix Scientific

039796-500MG
101848-204EA 193.85 USD
101848-204
2-(2-Benzoylhydrazino)-N-phenyl-2-thioxoacetamide
2-(2-Benzoylhydrazino)-N-phenyl-2-thioxoacetamide

Hazard: Irritant

Formula: C₁₅H₁₃N₃O₂S
MDL Number: MFCD12028066
CAS Number: 1134334-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR