2-Bromo-3-methoxypyridine-1-oxide

Supplier: Matrix Scientific

039792-500MG
101848-196EA 217.35 USD
101848-196
2-Bromo-3-methoxypyridine-1-oxide
2-Bromo-3-methoxypyridine-1-oxide

Hazard: Irritant

Formula: C₆H₆BrNO₂
MDL Number: MFCD09839263
CAS Number: 104819-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR