[4-Formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]acetic acid

Supplier: Matrix Scientific

039786-500MG
101848-184EA 338.46 USD
101848-184
[4-Formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]acetic acid
[4-Formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₄
MDL Number: MFCD05997906
CAS Number: 887408-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR