2-(4-Formyl-3-phenyl-1H-pyrazol-1-yl)-N-phenylacetamide

Supplier: Matrix Scientific

039785-500MG
101848-182EA 387.69 USD
101848-182
2-(4-Formyl-3-phenyl-1H-pyrazol-1-yl)-N-phenylacetamide
2-(4-Formyl-3-phenyl-1H-pyrazol-1-yl)-N-phenylacetamide

Hazard: Irritant

Formula: C₁₈H₁₅N₃O₂
MDL Number: MFCD06662332
CAS Number: 887407-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR