(4-Formyl-3-phenyl-1H-pyrazol-1-yl)acetic acid
Supplier: Matrix Scientific
039784-500MG
101848-180EA
307.69
USD
101848-180
(4-Formyl-3-phenyl-1H-pyrazol-1-yl)acetic acid
(4-Formyl-3-phenyl-1H-pyrazol-1-yl)acetic acid
Hazard: Irritant
Formula:
C₁₂H₁₀N₂O₃ |
MDL Number:
MFCD01160787 CAS Number: 353514-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...