2-(4-Aminophenoxy)-N-(4-chlorophenyl)acetamide

Supplier: Matrix Scientific

039783-500MG
101848-178EA 217.35 USD
101848-178
2-(4-Aminophenoxy)-N-(4-chlorophenyl)acetamide
2-(4-Aminophenoxy)-N-(4-chlorophenyl)acetamide

Hazard: Irritant

Formula: C₁₄H₁₃ClN₂O₂
MDL Number: MFCD06662308
CAS Number: 881607-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR