3-(2,3-Dimethylphenyl)-2-thioxo-1,3-thiazolidin-4-one

Supplier: Matrix Scientific

039781-500MG
101848-174EA 193.85 USD
101848-174
3-(2,3-Dimethylphenyl)-2-thioxo-1,3-thiazolidin-4-one
3-(2,3-Dimethylphenyl)-2-thioxo-1,3-thiazolidin-4-one

Hazard: Irritant

Formula: C₁₁H₁₁NOS₂
MDL Number: MFCD00175320
CAS Number: 108654-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR