2-Thioxo-3-[4-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one

Supplier: Matrix Scientific

039779-500MG
101848-170EA 217.35 USD
101848-170
2-Thioxo-3-[4-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one
2-Thioxo-3-[4-(trifluoromethyl)phenyl]-1,3-thiazolidin-4-one

Hazard: Irritant

Formula: C₁₀H₆F₃NOS₂
MDL Number: MFCD03184514
CAS Number: 57669-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR