Ethyl-2-hydroxy-2-(4-phenoxyphenyl)-3-phenylpropanoate
Supplier: Matrix Scientific
039777-500MG
101848-166EA
290.77
USD
101848-166
Ethyl-2-hydroxy-2-(4-phenoxyphenyl)-3-phenylpropanoate
Ethyl-2-hydroxy-2-(4-phenoxyphenyl)-3-phenylpropanoate
Hazard: Irritant
Formula:
C₂₃H₂₂O₄ |
MDL Number:
MFCD12028063 CAS Number: 1134334-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...