1-(4-Phenoxyphenyl)-2-phenylethanone

Supplier: Matrix Scientific

039776-500MG
101848-164EA 144.9 USD
101848-164
1-(4-Phenoxyphenyl)-2-phenylethanone
1-(4-Phenoxyphenyl)-2-phenylethanone

Hazard: Irritant

Formula: C₂₀H₁₆O₂
MDL Number: MFCD00156693
CAS Number: 3669-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR