2-(Carboxycarbonyl)benzoic acid

Supplier: Matrix Scientific

039773-500MG
101848-158EA 649.23 USD
101848-158
2-(Carboxycarbonyl)benzoic acid
2-(Carboxycarbonyl)benzoic acid

Hazard: Irritant

Formula: C₉H₆O₅
MDL Number: MFCD00090830
CAS Number: 528-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR