N,N-Dimethyl-1-(1H-pyrrol-2-yl)methanamine
Supplier: Matrix Scientific
Synonyms:
2-[(Dimethylamino)methyl]pyrrole
039771-500MG
101848-154EA
335.38
USD
101848-154
N,N-Dimethyl-1-(1H-pyrrol-2-yl)methanamine
N,N-Dimethyl-1-(1H-pyrrol-2-yl)methanamine
Hazard: Irritant
Formula:
C₇H₁₂N₂ MW: 124.19 g/mol |
MDL Number:
MFCD12028061 CAS Number: 14745-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...