Methyl [4-formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]acetate

Supplier: Matrix Scientific

039770-500MG
101848-152EA 217.35 USD
101848-152
Methyl [4-formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]acetate
Methyl [4-formyl-3-(4-methoxyphenyl)-1H-pyrazol-1-yl]acetate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₄
MDL Number: MFCD12028060
CAS Number: 1159697-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR