4-[(E)-2-(3-Fluorophenyl)vinyl]pyridine hydrochloride
Supplier: Matrix Scientific
039769-500MG
101848-150EA
193.85
USD
101848-150
4-[(E)-2-(3-Fluorophenyl)vinyl]pyridine hydrochloride
4-[(E)-2-(3-Fluorophenyl)vinyl]pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₁ClFN |
MDL Number:
MFCD12028059 CAS Number: 1185236-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...