1-(Pyridin-4-ylmethyl)pyrrolidin-2-one

Supplier: Matrix Scientific

039768-500MG
101848-148EA 335.38 USD
101848-148
1-(Pyridin-4-ylmethyl)pyrrolidin-2-one
1-(Pyridin-4-ylmethyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD12028058
CAS Number: 132312-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR