[(2-Methyltetrahydrofuran-2-yl)methyl]amine

Supplier: Matrix Scientific

039766-500MG
101848-144EA 289.8 USD
101848-144
[(2-Methyltetrahydrofuran-2-yl)methyl]amine
[(2-Methyltetrahydrofuran-2-yl)methyl]amine

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD09261457
CAS Number: 7179-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR