3-(1H-Imidazol-1-yl)propanamide

Supplier: Matrix Scientific

039765-500MG
101848-142EA 109.25 USD
101848-142
3-(1H-Imidazol-1-yl)propanamide
3-(1H-Imidazol-1-yl)propanamide

Hazard: Irritant

Formula: C₆H₉N₃O
MDL Number: MFCD02077567
CAS Number: 43115-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR