[1-(4-Methylbenzyl)-1H-imidazol-2-yl]methanol hydrochloride
Supplier: Matrix Scientific
039762-500MG
101848-136EA
387.69
USD
101848-136
[1-(4-Methylbenzyl)-1H-imidazol-2-yl]methanol hydrochloride
[1-(4-Methylbenzyl)-1H-imidazol-2-yl]methanol hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂O |
MDL Number:
MFCD12028056 CAS Number: 1185295-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...