2-(Ethylthio)pyrimidin-4-amine

Supplier: Matrix Scientific

039759-500MG
101848-130EA 387.69 USD
101848-130
2-(Ethylthio)pyrimidin-4-amine
2-(Ethylthio)pyrimidin-4-amine

Hazard: Irritant

Formula: C₆H₉N₃S
MDL Number: MFCD01646352
CAS Number: 54308-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR