2,2-Dimethyl-2,3-dihydro-1-benzofuran

Supplier: Matrix Scientific

039758-500MG
101848-128EA 358.46 USD
101848-128
2,2-Dimethyl-2,3-dihydro-1-benzofuran
2,2-Dimethyl-2,3-dihydro-1-benzofuran

Hazard: Irritant

Formula: C₁₀H₁₂O
MDL Number: MFCD12028054
CAS Number: 6337-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR