1-Benzyl-1H-1,2,3-triazol-5-amine

Supplier: Matrix Scientific

039757-500MG
101848-126EA 300 USD
101848-126
1-Benzyl-1H-1,2,3-triazol-5-amine
1-Benzyl-1H-1,2,3-triazol-5-amine

Hazard: Irritant

Formula: C₉H₁₀N₄
Melting Pt: 127…128 °C
MDL Number: MFCD11983439
CAS Number: 25784-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR