5-Amino-1-benzyl-1H-1,2,3-triazole-4-carboxylic acid

Supplier: Matrix Scientific

039756-500MG
101848-124EA 335.38 USD
101848-124
5-Amino-1-benzyl-1H-1,2,3-triazole-4-carboxylic acid
5-Amino-1-benzyl-1H-1,2,3-triazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₄O₂
MDL Number: MFCD02219534
CAS Number: 25784-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR