Methyl-3-methylisothiazole-4-carboxylate

Supplier: Matrix Scientific

039755-500MG
101848-122EA 362.25 USD
101848-122
Methyl-3-methylisothiazole-4-carboxylate
Methyl-3-methylisothiazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₂S
MDL Number: MFCD11983402
CAS Number: 74410-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR