2-Methoxy-3-(trifluoromethyl)benzoic acid
Supplier: Matrix Scientific
039752-500MG
101848-116EA
44.62
USD
101848-116
2-Methoxy-3-(trifluoromethyl)benzoic acid
2-Methoxy-3-(trifluoromethyl)benzoic acid
Hazard: Irritant
Formula:
C₉H₇F₃O₃ |
MDL Number:
MFCD09832311 CAS Number: 937068-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...