5-Bromo-1-methyl-1H-indole-3-carbaldehyde

Supplier: Matrix Scientific

039748-500MG
101848-106EA 338.46 USD
101848-106
5-Bromo-1-methyl-1H-indole-3-carbaldehyde
5-Bromo-1-methyl-1H-indole-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₈BrNO
MDL Number: MFCD03906373
CAS Number: 10102-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR