5-Nitro-1H-pyrazole-3-carboxamide
Supplier: Matrix Scientific
039745-500MG
101848-100EA
387.69
USD
101848-100
5-Nitro-1H-pyrazole-3-carboxamide
5-Nitro-1H-pyrazole-3-carboxamide
Hazard: Irritant
Formula:
C₄H₄N₄O₃ |
MDL Number:
MFCD12028052 CAS Number: 297149-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...