Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)-butanoic acid

Supplier: Matrix Scientific


039744-500MG
101848-098EA 247.15 USD InStock
101848-098
4-(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)-butanoic acid
4-(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)butanoic acid
CAS: 898050-13-0
MDL Number: MFCD03410243
Molecular Formula: C9H13N3O4
Molecular Weight: 227,22
Hazard: Irritant


Formula: C₉H₁₃N₃O₄
MW: 227,22 g/mol
MDL Number: MFCD03410243
CAS Number: 898050-13-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty